(SP-4-4)-[4-(Diphenylphosphino-κP)-6-(diphenylphosphino)-10H-phenoxazine](methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC]palladium 98%

Supplier: Ambeed
Warning

A1155290-250MG A1155290-1G A1155290-5G
77549-284EA 218 USD
77549-284 77566-070 77866-056
(SP-4-4)-[4-(Diphenylphosphino-κP)-6-(diphenylphosphino)-10H-phenoxazine](methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC]palladium 98%
(SP-4-4)-[4-(Diphenylphosphino-κP)-6-(diphenylphosphino)-10H-phenoxazine](methanesulfonato-κO)[2'-(methylamino-κN)[1,1'-biphenyl]-2-yl-κC]palladium

Precaución: For research use only.

Formula: C₅₀H₄₂N₂O₄P₂PdS
MDL Number: MFCD30718059
CAS Number: 1878105-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR