Ru(II)-(S)-Pheox ≥97%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(acetonitrile)[2-[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl-N]phenyl]ruthenium(II) Hexafluorophosphate, Ruthenium-tetrakis(acetonitrile)[2-[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl-κN3]phenyl-κC]-hexafluorophosphate

A1155271-1G A1155271-250MG A1155271-100MG
77549-282EA 238.96 USD
77549-282 77570-926 77592-264
Ru(II)-(S)-Pheox ≥97%
Ru(II)-(S)-Pheox

Precaución: for Research Use Only

Formula: C₂₃H₂₄F₆N₅OPRu
MW: 632.51 g/mol
MDL Number: MFCD31618155
CAS Number: 1259070-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR