3-Hydroxy-2,2-bis(hydroxymethyl)propyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate ≥97%

Supplier: Ambeed
Warning

A1151859-100MG A1151859-1G
77549-270EA 85.49 USD
77549-270 77570-890
3-Hydroxy-2,2-bis(hydroxymethyl)propyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate ≥97%
3-Hydroxy-2,2-bis(hydroxymethyl)propyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate

Precaución: for Research Use Only

Formula: C₂₂H₃₆O₆
MW: 396.517 g/mol
MDL Number: MFCD32206581
CAS Number: 26347-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR