[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][[2'-(diphenylphosphino)[1,1'-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)palladium 98%

Supplier: Ambeed
Warning

A1150979-5G A1150979-1G A1150979-250MG A1150979-10G
77549-266EA 122 USD
77549-266 77566-056 77570-884 77783-313
[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][[2'-(diphenylphosphino)[1,1'-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)palladium 98%
[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][[2'-(diphenylphosphino)[1,1'-binaphthalen]-2-yl]diphenylphosphine-κP](methanesulfonato-κO)palladium

Precaución: For research use only.

Formula: C₅₇H₄₅NO₃P₂PdS
MDL Number: MFCD28144629
CAS Number: 2151915-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR