4-Bromo-3-chloro-1-methylpyridin-2(1H)-one ≥97%

Supplier: Ambeed
Warning

A1146632-1G A1146632-250MG A1146632-5G
77549-250EA 307 USD
77549-250 77865-899 77865-900
4-Bromo-3-chloro-1-methylpyridin-2(1H)-one ≥97%
4-Bromo-3-chloro-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MDL Number: MFCD32701974
CAS Number: 2172654-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR