3-(2-(2-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethoxy)ethoxy)ethoxy)propanoic acid ≥95%

Supplier: Ambeed
Warning

A1146494-250MG A1146494-1G A1146494-100MG
77549-248EA 341 USD
77549-248 77570-840 77619-998
3-(2-(2-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethoxy)ethoxy)ethoxy)propanoic acid ≥95%
3-(2-(2-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethoxy)ethoxy)ethoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₇N₃O₉
MDL Number: MFCD32173779
CAS Number: 2138440-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR