3-Bromopropane-1-sulfonamide ≥95%

Supplier: Ambeed
Warning

A1134183-250MG A1134183-100MG
77549-204EA 239.99 USD
77549-204 77595-756
3-Bromopropane-1-sulfonamide ≥95%
3-Bromopropane-1-sulfonamide

Precaución: for Research Use Only

Formula: C₃H₈BrNO₂S
MW: 202.0701 g/mol
MDL Number: MFCD19382021
CAS Number: 88496-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR