tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 1-BOC-7-Azaindole-3-boronic acid, pinacol ester

A112305-1G A112305-250MG A112305-100MG A112305-10G
77549-182EA 55 USD
77549-182 77565-970 77880-904 77880-905
tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate 98%
tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Precaución: For research use only.

Formula: C₁₈H₂₅BN₂O₄
MDL Number: MFCD12407268
CAS Number: 942070-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR