4-Cyclohexylbenzenesulfonyl chloride ≥97%

Supplier: Ambeed
Danger

Synonyms: 4-Cyclohexylbenzene-1-sulphonyl chloride

A112021-5G A112021-1G
77549-176EA 67.98 USD
77549-176 77570-670
4-Cyclohexylbenzenesulfonyl chloride ≥97%
4-Cyclohexylbenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₂S
MW: 258.76 g/mol
MDL Number: MFCD00625713
CAS Number: 56354-57-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR