(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-3,3-dimethylazetidine-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A110721-1G A110721-100MG
77549-144EA 659.2 USD
77549-144 77747-817
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-3,3-dimethylazetidine-2-carboxylic acid ≥98%
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-3,3-dimethylazetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MW: 351.395 g/mol
MDL Number: MFCD31705408
CAS Number: 2231665-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR