4-Acetylpyridine 1-oxide ≥98%

Supplier: Ambeed
Warning

A109426-1G A109426-250MG
77549-106EA 86 USD
77549-106 77549-108
4-Acetylpyridine 1-oxide ≥98%
4-Acetylpyridine 1-oxide

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MDL Number: MFCD00955960
CAS Number: 2402-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR