2-Formyl-3-hydroxybenzoic acid ≥95%

Supplier: Ambeed
Warning

A1085368-100MG A1085368-250MG
77549-092EA 144 USD
77549-092 77592-168
2-Formyl-3-hydroxybenzoic acid ≥95%
2-Formyl-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆O₄
MDL Number: MFCD16999703
CAS Number: 1243461-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR