5-Bromo-4,6-dimethylpyridin-3-amine ≥98%

Supplier: Ambeed
Warning

A1075315-1G A1075315-100MG A1075315-250MG
77549-072EA 200 USD
77549-072 77783-114 77783-115
5-Bromo-4,6-dimethylpyridin-3-amine ≥98%
5-Bromo-4,6-dimethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD07439966
CAS Number: 890092-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR