2-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)ethan-1-amine ≥97%

Supplier: Ambeed
Danger

A107248-100MG A107248-250MG A107248-50MG A107248-1G
77549-056EA 233.81 USD
77549-056 77549-058 77549-060 77570-434
2-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)ethan-1-amine ≥97%
2-(3-(But-3-yn-1-yl)-3H-diazirin-3-yl)ethan-1-amine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃
MW: 137.18 g/mol
MDL Number: MFCD29918288
CAS Number: 1450752-97-2
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR