rel-N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(1-(pyridin-2-yl)methanimine) ≥98%

Supplier: Ambeed
Warning

A1059600-1G A1059600-100MG A1059600-250MG
77549-022EA 548 USD
77549-022 77570-342 77570-344
rel-N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(1-(pyridin-2-yl)methanimine) ≥98%
rel-N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(1-(pyridin-2-yl)methanimine)

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₄
MW: 292.38 g/mol
CAS Number: 284497-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR