tert-Butyl hydrogen malonate ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-Tert-butoxy-3-oxopropanoic acid, mono-tert-Butyl malonate

A104350-100G A104350-25G
77548-988EA 45 USD
77548-988 77570-264
tert-Butyl hydrogen malonate ≥97%
tert-Butyl hydrogen malonate

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MDL Number: MFCD00191886
CAS Number: 40052-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR