(±)-3-Undecanol ≥98%

Supplier: Ambeed
Danger

Synonyms: (±)-3-Hydroxy undecane, 3-Undecanol

A104163-25G A104163-5G A104163-1G
77548-984EA 142.14 USD
77548-984 77570-252 77623-858
(±)-3-Undecanol ≥98%
(±)-3-Undecanol

Precaución: for Research Use Only

Formula: C₁₁H₂₄O
MW: 172.31 g/mol
MDL Number: MFCD00046730
CAS Number: 6929-08-4
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR