5-Iodopent-1-yne ≥97%

Supplier: Ambeed
Danger

Synonyms: 5-Iodo-1-pentyne

A104104-5G A104104-1G A104104-250MG A104104-25G
77548-982EA 206 USD
77548-982 77570-250 77610-804 77682-954
5-Iodopent-1-yne ≥97%
5-Iodopent-1-yne

Precaución: For research use only.

Formula: C₅H₇I
MW: 194.01 g/mol
MDL Number: MFCD09907724
CAS Number: 2468-55-5
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR