tert-Butyl-4-formylpiperidine-1-carboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-formylpiperidine-1-carboxylate, N-Boc-4-Formylpiperidine, 1-BOC-4-Piperidinecarboxaldehyde, 1-Boc-4-formylpiperidine, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxaldehyde, 1-(tert-Butoxycarbonyl)-4-formylpiperidine, 1-Boc-4-piperidinecarboxaldehyde, Tert-butyl 4-formylpiperidine-1-carboxylate

A103974-100G A103974-10G A103974-25G A103974-500G
77548-972EA 60.77 USD
77548-972 77548-974 77548-976 77570-232
tert-Butyl-4-formylpiperidine-1-carboxylate ≥97%
tert-Butyl-4-formylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.28 g/mol
MDL Number: MFCD02179019
CAS Number: 137076-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR