Fluoromethanesulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A1039480-5G A1039480-100MG A1039480-1G
77548-970EA 3707 USD
77548-970 77782-956 77782-957
Fluoromethanesulfonyl chloride ≥95%
Fluoromethanesulfonyl chloride

Precaución: for Research Use Only

Formula: CH₂ClFO₂S
MDL Number: MFCD19200094
CAS Number: 42497-69-8
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR