3-(Dimethylamino)propane-1-thiol ≥95%

Supplier: Ambeed
Danger

A1037655-1G A1037655-5G
77548-964EA 109 USD
77548-964 77570-218
3-(Dimethylamino)propane-1-thiol ≥95%
3-(Dimethylamino)propane-1-thiol

Precaución: for Research Use Only

Formula: C₅H₁₃NS
MDL Number: MFCD00128221
CAS Number: 42302-17-0
UN: 1228
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR