tert-Butyl-2-(aminomethyl)phenylcarbamate ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)aniline, 1-BOC protected

A103739-1G A103739-5G
77548-962EA 79.31 USD
77548-962 77666-496
tert-Butyl-2-(aminomethyl)phenylcarbamate ≥97%
tert-Butyl-2-(aminomethyl)phenylcarbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD04037902
CAS Number: 849020-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR