6-Bromohex-1-yne ≥95%

Supplier: Ambeed
Warning

A103602-1G A103602-100MG A103602-250MG
77548-958EA 625.21 USD
77548-958 77610-788 77610-790
6-Bromohex-1-yne ≥95%
6-Bromohex-1-yne

Precaución: for Research Use Only

Formula: C₆H₉Br
MW: 161.04 g/mol
MDL Number: MFCD02258668
CAS Number: 66977-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR