2,6-Bis(trifluoromethyl)benzoic acid 98

Supplier: Ambeed
Warning

A101865-5G A101865-1G A101865-25G
77548-920EA 62 USD
77548-920 77876-785 77876-786
2,6-Bis(trifluoromethyl)benzoic acid 98
2,6-Bis(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₄F₆O₂
MW: 258.12
Boiling Pt: 260
MDL Number: MFCD00000376
CAS Number: 24821-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR