Acetobromo-α-D-glucuronic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: BROMOMETHYLTRIACETYL GLUCURONATE, Acetobromo-α-D-glucuronic Acid Methyl Ester, Methyl acetobromo-α-D-glucuronate, (2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

A101618-100G A101618-25G A101618-10G A101618-500G
77548-912EA 243 USD
77548-912 77548-914 77565-762 77570-110
Acetobromo-α-D-glucuronic acid methyl ester 97%
Acetobromo-alpha-D-glucuronic acid methyl ester

Precaución: For research use only.

Formula: C₁₃H₁₇BrO₉
MDL Number: MFCD00061613
CAS Number: 21085-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR