Cyclopropanamine hydrochloride ≥97%

Supplier: Ambeed
Warning

A1003677-1G A1003677-5G A1003677-25G
77548-872EA 36 USD
77548-872 77548-874 77570-024
Cyclopropanamine hydrochloride ≥97%
Cyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClN
MW: 93.5553 g/mol
MDL Number: MFCD20483529
CAS Number: 18600-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR