(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-Hse(Me)-OH
A1002983-1G
A1002983-5G
77548-864EA
279.13
USD
77548-864
77570-014
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid ≥98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid
Precaución: for Research Use Only
Formula:
C₂₀H₂₁NO₅ MW: 355.3845 g/mol |
MDL Number:
MFCD07366856 CAS Number: 1272755-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...