Cell Culture Flasks
Supplier: NEST SCIENTIFIC
707001 707003 707011 707013 708001 708003 708011 708013 709001 709003 709011 709013 720001 720003 720011 720013 721001 721003 721011 721013
77508-556CS 181.24 USD
77508-556 77508-558 77508-560 77508-562 77508-564 77508-566 77508-568 77508-570 77508-572 77508-574 77508-576 77508-578 77508-612 77508-614 77508-616 77508-618 77508-620 77508-622 77508-624 77508-626
Cell Culture Flasks
Flasks Cell Culture Flasks
Cell culture flasks are available in cell growth areas ranging from 25 to 225 cm². Tissue culture treated or non-treated flasks available with a vent cap or plug seal cap to meet requirements. Vacuum plasma tissue culture treated provides optimum cell adherence. Non-treated flasks utilize yellow caps to be different from TC flasks which have teal caps. They are for suspension cell growth.
  • Made of high clarity, 100% virgin polystyrene
  • Sterilized by E-beam, SAL=10⁻⁶
  • Non-Pyrogenic, DNase/Rnase free
  • Frosted writing and clear graduations
  • Notched bottom for slip free stacking
Clear lot number for batch traceability.
Vent caps with 0.22 μm hydrophobic filters to ensure gas exchange without contamination.
Envase: Packaged in sterile, zip-sealable bags.