Sterile Swabs
Supplier: NEST SCIENTIFIC
202003 202004
77507-424CS 1030 USD
77507-424 77507-426
Sterile Swabs
Swabs Swabs for Sampling
Nest nylon flocked swabs provide optimum specimen collection because of their ability to collect more specimen at the collection site and to more rapidly release the entire sample specimen.
  • Easy to operate and ready to use
  • The preservation solution poses a function of inactivation
  • Disposable
  • Non toxic
  • Individually wrapped and sterile
The product is intended for sample collection, transportation, storage and more.
Certificaciones: ISO 9001 and 13485 certified, USP 6.