Defender™ 6000 Industrial Washdown Bench Scales
Supplier: Ohaus
30631706 30631707 30631718 30631719 30575563 30575564 30575565 30575566 30575567 30631720 30631721 30631722 30631723 30631724
77435-088EA 2232.74 USD
77435-088 77435-090 77435-092 77435-094 77435-038 77435-040 77435-042 77435-044 77435-046 77435-096 77435-098 77435-100 77435-102 77435-104
Defender™ 6000 Industrial Washdown Bench Scales
Balances and Scales
Advanced IP68/IP69k washdown bench scales. The durable Defender 6000 stands up to harsh cleaning in processing and packaging environments with IP 68 immersion/ IP69K washdown protection, 316 stainless steel base and hermetically-sealed load cell.
  • Supports HACCP systems with a 316 stainless steel base and sturdy indicator providing corrosion protection in food, chemical processing and packaging
  • Versatile scales that include fast weight display and multiple software modes
The Defender 6000 series extreme washdown bench scale is specifically designed for food, chemical and pharmaceutical applications, including production and packaging. Defender 6000 handles the most demanding applications with 316 stainless steel platform and frame, and hermetically-sealed stainless steel IP69K load cell. Defender 6000 meets metrological approvals, NSF listing, and supporting HACCP systems.
Certificaciones: Defender 6000 is NSF certified.