Rubbermaid® Commercial Invader® Side-Gate Wet-Mop Handle

Supplier: Janitorial Supplies
RCPH146 RCPH136 RCPH126
89073-444EA 111.88 USD
89073-444 89073-442 89073-436 75778-008
Rubbermaid® Commercial Invader® Side-Gate Wet-Mop Handle
Mops
Solvent- and acid-resistant frame enables use with all commercial floor stripping agents.

  • Easily remove and replace mop heads
  • Side gate makes wet mop head connection easy
  • Solvent and acid-resistant frame
  • For use with 1" headband mop heads

Easy-change, side-gate design allows you to quickly slide mop heads on or off. Adjustable thumb wheel enables stress-free, tight and secure clamp down.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR