Uvex Flex Seal® Safety Goggles, Honeywell Safety
Supplier: Honeywell Safety Products
S3400X S3410X
33002-546CS 1877.24 USD
33002-546 33002-551 33002-548
Uvex Flex Seal® Safety Goggles, Honeywell Safety
Goggles
Lightweight silicone body conforms to the face to protect against impacts and chemical splashes. The silicone is cured to stay soft and supple even in extreme temperatures.
  • Provide excellent impact and chemical-splash resistance
  • Fit over most prescription eyewear
Low-profile and optically correct clear polycarbonate lens provides the full field of vision. Uvextreme® antifog lens coating and indirect venting system minimize fogging. Goggles fit over most prescription eyewear and feature pivoting clips that permit the wearer to position them on a hard hat or position the headband above the ears. Frame color is navy.
Certificaciones: Goggles meet ANSI Z87+ and CSA Z94.3 standards.