Color Mono II Test Kits, ASI

Supplier: ARLINGTON SCIENTIFIC INC.

450100 450050 4501000 450025
80087-378EA 396.32 USD
80087-378 80087-382 80087-376 80087-380
Color Mono II Test Kits, ASI
Clinical Diagnostic Supplies Clinical Reagents and Kits

The ASI Color Mono II Test is a convenient and specific agglutination test for the qualitative and semiquantitative detection of heterophil antibodies in human serum or plasma associated with infectious mononucleosis (IM). No initial dilution of patient sample is required for this test.


  • Easy to read results in 2 minutes
  • Color enhanced agglutination test
  • Qualitative and semiquantitative procedures
  • All testing materials needed to perform the test included in each kit
  • Can be used with serum or plasma
  • Color-coded reactive and nonreactive controls with dropper tip vials included in the kit.
  • Storage requirement: 2-8ºC
  • Proudly made in USA
  • Longest shelf life - up to 24 Months
  • Proven by AAB, API, and CAP surveys
  • Sensitivity 1:28 Davidsohn titer
  • Specificity >95%


Certificaciones: CE marked. CPT Code 86308. 510(k)-K821386

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR