Calcium chloride

Supplier: RPI
Warning

C20010-2500.0 C20010-500.0
77510-188EA 283.37 USD
77510-188 77510-190
Calcium chloride
Calcium chloride
Formula: CaCl₂
Melting Pt: 772 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now

Specification Test Results

Appearance White crystalline powder
Assay (Titration) ≥ 96.0 %
Titrable Base ≤ 0.006 meq/g
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR