BetaBasic™ 8 Javelin Guard Columns
Supplier: Thermo Scientific
71405-012106 71405-022106
10043-300EA 685.77 USD
10043-300 10043-306
BetaBasic™ 8 Javelin Guard Columns
Chromatography Columns
BetaBasic™ 8 Javelin guard columns are a good first choice for compounds typically considered difficult to analyze. It is used for analytical HPLC Column.
  • Has a carbon load of 7%
  • pH 2 to 8
  • L7 USP Type
  • 200 m²/g surface area
  • Not encapped
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement