Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 20–25, Non sterile, Aspen Surgical
Supplier: Aspen Surgical Products
371325 371320 371321 371322 371323
89176-378CS 167.39 USD
89176-378 25860-020 25860-042 25860-064 25860-086
Bard-Parker® Rib-Back™ Carbon Steel Scalpel Blades, Nos. 20–25, Non sterile, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Blades
For use with No
4 and No. 6 Bard-Parker® scalpel handles. Fine blade carbon steel. Rib-Back™ is designed for greater cutting control and strength. Specially ground and carefully inspected. Blades individually packed in foil pouch.