Adenine

Supplier: RPI
Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

A11500-100.0 A11500-1000.0 A11500-25.0 A11500-5.0 A11500-500.0
76344-584EA 259.52 USD
76344-584 76344-586 76344-588 77510-158 77510-160
Adenine
Adenine

A nucleobase with a variety of roles in biochemistry including cellular respiration, in the form of both the energy-rich adenosine triphosphate and the cofactors nicotinamide adenine dicucleotide and flavin adenine dinucleotide, and protein synthesis, as a chemical component of DNA and RNA.


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: >350 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00041790
CAS Number: 73-24-5
Merck Index: 13,00152

Order Now

Specification Test Results

Appearance White to almost white powder almost white to slightly yellowish crystalline powder
Identification (IR) Conforms to reference
Identification (Retention Time) Conforms to retention time of reference solution
Assay (HPLC, Dry Basis) 98.0 - 102.0 %
Loss on Drying 110 °C for 4Hrs ≤ 1.0 %
Residue on Ignition ≤ 0.1 %
Related Compounds (Individual Impurities) ≤ 0.1 %
Related Compounds (Total Impurities) ≤ 2.0 %
Heavy Metals (as Pb) ≤ 10 ppm
Total Aerobic Microbial Count (TAMC) ≤ 10 cfu/g
Total Combined Yeast and Mold Count ≤ 10 cfu/g
BSE/TSE Free BSE/TSE Free
Non Animal Origin Non Animal Origin
Grade USP
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR