Bisacrylamide
Supplier: RPI
|
Warning
|
A11270-100.0
A11270-1000.0
A11270-25.0
A11270-500.0
A11270-250.0
76344-540EA
85.85
USD
76344-540
76344-542
76344-544
76344-546
77510-156
Bisacrylamide
Bisacrylamide
Formula:
(H₂C=CHCONH)₂CH₂ |
MDL Number:
MFCD00008625 CAS Number: 110-26-9 |
Specification Test Results
Appearance | White Crystalline Powder |
Absorbance 290 nm (1% water ) | ≤ 0.2 |
Acrylic Acid | ≤ 0.001 % |
Conductivity (2% Water) | ≤ 5 umhos |
DNase | Not Detected |
Identification | Pass |
pH (1%, 0.1M NaCl) 25 °C | ≥ 5.0 |
Protease | Not Detected |
Purity | ≥ 99.0 % |
RNase | Not Detected |
BSE/TSE Free | BSE/TSE Free |
Country of Origin | Report |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...