Blood Bank Saline Solutions, Azer Scientific
Supplier: AZER SCIENTIFIC INC
ES124320L ES124310L ES124420L ES12444G ES124410L
16005-094EA 67.5 USD
16005-094 16005-096 16005-088 16005-092 16005-090
Blood Bank Saline Solutions, Azer Scientific
Buffers Cell Culture Buffers
Blood bank saline is pH-controlled for a pH between 6.5 and 7.0
  • Meets NCCLS standard ASI/BB-1
  • Both standard and buffered saline remain stable without preservatives
  • Cubes feature a spigot for easy dispensing
Buffered saline is tightly controlled to a pH range of 7.0 to 7.2 and is specially designed to enhance weak agglutination reactions in blood bank procedures.