Coomassie® brilliant blue R-250

Supplier: RPI
Warning

Synonyms: Serva Blue R, Brillant blue R, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1), Coomassie Blue R 250, Brilliant Cyanine 6B, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, Acid Blue 83, Brilliant Blue R250, Brilliant Blue R, Brilliant indocyanine 6B

C25250-100.0 C25250-25.0 C25250-5.0 C25250-500.0
77510-194EA 469.14 USD
77510-194 77510-196 77510-198 77510-200
Coomassie® brilliant blue R-250
Coomassie® brilliant blue R-250
Formula: C₄₅H₄₄N₃NaO₇S₂
MW: 825.98 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00041762
CAS Number: 6104-59-2

Order Now

Specification Test Results

Appearance Blue to red crystalline powder
Em (550 nm, Water, Dry Basis) ≥33000
Protein Staining Pass
Country of Origin Report

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR