Guanidinium thiocyanate

Supplier: RPI
Danger

G54250-4000.0 G54250-500.0 G54250-5000.0 G54250-1000.0
77510-272EA 1051.16 USD
77510-272 77510-274 77510-276 77510-270
Guanidinium thiocyanate
Guanidinium thiocyanate

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: CH₅N₃·CHNS
Storage Temperature: Room Temperature
MDL Number: MFCD00013027
CAS Number: 593-84-0
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless liquid
Insoluble Matter Passes Filter Test
pH @25 °C 4.5 - 7.0
Refractive Index 1.474 - 1.483
Comment Crystallized product can be dissolved again by slightly heating to 40-50 °C.
Country of Origin US


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR