Dextran 70

Supplier: RPI

D30850-100.0 D30850-1000.0 D30850-10000.0 D30850-500.0 D30850-5000.0
77510-240EA 138.12 USD
77510-240 77510-242 77510-244 77510-246 77510-248
Dextran 70
Dextran

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: (C₆H₁₀O₅)n
Storage Temperature: Refrigerator
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now

Specification Test Results

Appearance White to slightly off white powder
Loss on Drying ≤ 7.0 %
Ash Content ≤ 2.0 %
Specific Rotation[α ]20D (2% solution) +195 - +203 °
Intrinsic Viscosity @ 37 oC (1% Solution) 0.22 - 0.27
pH (6% Solution) 4.5 - 7.0
Color of solution @ 375nm (6% solution) ≤ 0.15 OD
Cold water solubility (1% solution) Soluble in cold water
Non Animal Origin Conforms
BSE/TSE Free BSE/TSE Free
Country of Origin CA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR