Calbiochem® Caspase-3 Inhibitor III
Supplier: MilliporeSigma
218750-1MG 218750-5MG
80050-812EA 113.76 USD
80050-812 80050-814
Calbiochem® Caspase-3 Inhibitor III
Proteins and Peptides
A potent, cell-permeable, and irreversible inhibitor of caspase-3 as well as caspase-6, caspase-7, caspase-8, and caspase-10.