Saccomanno Fluid, Epredia™
Supplier: Epredia
76160 76161
84000-146CS 993.32 USD
84000-146 84000-148
Saccomanno Fluid, Epredia™
Stains Microscopy and staining reagents
Store specimens for weeks with Epredia™ Saccomanno Fluid, which acts as an excellent general cytological fixative.
  • Prevents air drying artifacts
  • For upper respiratory tract cytology, most notably sputum
  • Each prefilled container is labeled with a do not drink warning
  • Clear containers and tinted solution make specimen identification easy
  • Ready-to-use cytology fixative is an alcohol and polyethylene glycol (Carbowax™) solution
The ready-to-use fixative is a mixture of alcohol and polyethylene glycol that is suited especially for upper respiratory tract cytology, particularly sputum. The fluid permits storage of specimens for weeks and prevents air drying artifacts from appearing in the samples. It can also serve as a good general cytology fixative. The solution is tinted for easy specimen identification. The poly containers are clear and labeled with a 'Do Not Drink' warning, since specimens may be collected at home.