Neutralyde™ Formalin Neutralizing Liquid, Azer Scientific
Supplier: VWR International
ES767 ES766
95039-816EA 233.78 USD
95039-816 95039-814
Neutralyde™ Formalin Neutralizing Liquid, Azer Scientific
Spill Control Materials
Neutralyde™ Formalin Neutralizing Liquid is designed to safely neutralize and dispose of formalin in the laboratory environment without the threat of exposure to hazardous vapors.
  • No pH adjustment required
  • Phosphate-free
  • Complete neutralization in eight hours
  • One gallon of Neutralyde for five gallons of 10% formalin waste
  • Effective on formaldehyde and glutaraldehyde solutions
  • Safe and gentle reaction
A single gallon of Neutralyde liquid treats up to five gallons of 10% formalin waste and provides a safe and gentle reaction with virtually no risk to personnel.
Neutralyde™ is a phosphate-free, eco-friendly liquid neutralizer that is effective on formaldehyde and glutaraldehyde solutions. Treated waster may be disposed of in sanitary drain.
Información para pedidos: Odering: Available in 1, 2 1/2 or 5 gallon cubes
Envase: Packaged in cubitainers equipped with a dispensing spigot.