Accessories for Compresstome® Specimen Tube Kits for VF-510-0Z
Supplier: PRECISIONARY INSTRUMENTS LLC
VFBHSK15.5510 VFBHSKVM20510 VFSPSK51015.5 VFSPSK51020
77435-400EA 1182.72 USD
77435-400 77435-410 77435-482 77435-484
Accessories for Compresstome® Specimen Tube Kits for VF-510-0Z
Microtomes and Microtome Assemblies Microtomes
Compresstome Specimen Tube Kits are compatible with the Compresstome VF-510-0Z.
  • Specimen tube kits are available in 15.5 or 20 mm diameters
Compresstome specimen tube kits with 15.5 or 20 mm diameter include two specimen tubes with matching plungers, one chilling block, and one buffer tray.
Compresstome specimen tube kits with 15 or 20 mm diameter accessories and a blade holder include two specimen tubes with matching plungers, one chilling block, one buffer tray, and one blade holder.