These models feature a TwinGuard Eco cooling refrigeration system for redundant sample protection, a microprocessor controller and digital display providing great visibility and intuitive operation. The models are equipped with three access ports and an USB port on the control panel for data log retrieval. Door glass heating for condensation prevention on the glass.
- Utilizes Hydrocarbon [HC] refrigerants that are compliant with US EPA SNAP legislation for environmental sustainability
- Digital display control panel that allows convenient but secure user control. Refrigerator temperature can be displayed in 0.1 °C increments USB data download available
- Audible and visual alarms with adjustable set points warn of deviations from set point temperatures
- Condensation prevention by way of warm air emissions across the glass
- Choice of models with wire shelves or wire roll out drawers
Natural refrigerants, variable speed inverter compressors and integrated electronics combine to help achieve facility sustainability objectives by minimizing environmental impact.
Certificaciones: Energy Star Certified, QPS Listed.
Precaución: Please note the voltage requirements are: 115 V/60 Hz/single phase/15-amp breaker.