2-(Pyridin-2-yldisulfanyl)ethanol ≥97%

Supplier: Ambeed
Danger

Synonyms: (2-pyridyldithio)-PEG1-hydrazine

A1001691-1G A1001691-5G A1001691-250MG A1001691-100MG
77548-858EA 78 USD
77548-858 77548-860 77570-002 77619-478
2-(Pyridin-2-yldisulfanyl)ethanol ≥97%
2-(Pyridin-2-yldisulfanyl)ethanol

Precaución: for Research Use Only

Formula: C₇H₉NOS₂
MDL Number: MFCD22551638
CAS Number: 111625-28-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR