3-Bromo-1-cyclopropyl-1H-pyrazole ≥97%

Supplier: Ambeed
Warning

A1001563-1G A1001563-250MG A1001563-5G
77548-856EA 310 USD
77548-856 77570-000 77782-688
3-Bromo-1-cyclopropyl-1H-pyrazole ≥97%
3-Bromo-1-cyclopropyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MDL Number: MFCD28054693
CAS Number: 1622840-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR