Formaldehyde 37%, VWR®
Supplier: VWR International
|
Danger
|
Stabilization:
Stabilized with methanol
VWR®, part of Avantor has industry leading Anatomical Pathology reagents available for your lab.
- ISO 13485 Certified
- F-style gallons for easier storage and fewer spills
- Transparency and availability to help you with reliability
Certificaciones: ISO 13485 Certified.
Información de suministro: Supplied with screw cap and pouring ring.
Envase: The bottles are packaged in shrinked-wrapped modules.
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 1198 ADR: 3 (8),III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...